COMH93D5DB95 Combi-Blocks, Inc.

7-Methyl-1h-pyrazolo[4,3-b]pyridine

  • MDL number MFCD12196962
  • Cas Number : 1130309-70-4
  • Molecular Weight : 133.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3428

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3428
Availability details for
Quantity
  • Availability
  • Add to Cart