COMH93D5DBD2 Combi-Blocks, Inc.

2H-3,4'-Bi-1,2,4-triazole-5-carboxylic acid

  • MDL number MFCD12402407
  • Cas Number : 842977-15-5
  • Molecular Weight : 180.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3503

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3503
Availability details for
Quantity
  • Availability
  • Add to Cart