COMH93D5DBF8 Combi-Blocks, Inc.

3-(2-(tert-Butoxycarbonyl)-1,2,3,4-tetrahydroisoquinolin-6-yl)propanoic acid

  • MDL number MFCD12406216
  • Cas Number : 1158756-02-5
  • Molecular Weight : 305.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3553

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3553
Availability details for
Quantity
  • Availability
  • Add to Cart