COMH93D5DC25 Combi-Blocks, Inc.

4-Bromo-n-(oxan-4-yl)benzene-1-sulfonamide

  • MDL number MFCD12524249
  • Cas Number : 1183706-81-1
  • Molecular Weight : 320.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3605

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3605
Availability details for
Quantity
  • Availability
  • Add to Cart