COMH93D5DC97 Combi-Blocks, Inc.

(S)-1-(Pyridin-2-yl)propan-1-amine hydrochloride

  • MDL number MFCD12759573
  • Cas Number : 339312-61-7
  • Molecular Weight : 172.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3746

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3746
Availability details for
Quantity
  • Availability
  • Add to Cart