COMH93D5DDBD Combi-Blocks, Inc.

(3S,4S)-Ethyl 4-methylpyrrolidine-3-carboxylate

  • MDL number MFCD13152225
  • Cas Number : 1260603-17-5
  • Molecular Weight : 157.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4103

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4103
Availability details for
Quantity
  • Availability
  • Add to Cart