COMH93D5DDE8 Combi-Blocks, Inc.

6-Fluoro-3-iodo-1h-indole

  • MDL number MFCD13177480
  • Cas Number : 741265-04-3
  • Molecular Weight : 261.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4157

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4157
Availability details for
Quantity
  • Availability
  • Add to Cart