COMH93D5DE96 Combi-Blocks, Inc.

5-(Chloromethyl)-2-methyloxazole

  • MDL number MFCD13190299
  • Cas Number : 1196146-69-6
  • Molecular Weight : 131.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4369

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4369
Availability details for
Quantity
  • Availability
  • Add to Cart