COMH93D5DEAF Combi-Blocks, Inc.

5-Fluorobenzofuran-3-carbaldehyde

  • MDL number MFCD13191785
  • Cas Number : 721943-19-7
  • Molecular Weight : 164.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4397

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4397
Availability details for
Quantity
  • Availability
  • Add to Cart