COMH93D5DEB3 Combi-Blocks, Inc.

3,5-Difluoro-2-methylaniline

  • MDL number MFCD13191858
  • Cas Number : 1092350-38-3
  • Molecular Weight : 143.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4401

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4401
Availability details for
Quantity
  • Availability
  • Add to Cart