COMH93D5DEBD Combi-Blocks, Inc.

2,3-Difluoro-6-iodobenzonitrile

  • MDL number MFCD13191970
  • Cas Number : 935660-92-7
  • Molecular Weight : 265

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4413

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4413
Availability details for
Quantity
  • Availability
  • Add to Cart