COMH93D5DECE Combi-Blocks, Inc.

4'-Bromo-2',3'-difluoroacetophenone

  • MDL number MFCD13192197
  • Cas Number : 1007346-28-2
  • Molecular Weight : 235.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4432

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4432
Availability details for
Quantity
  • Availability
  • Add to Cart