COMH93D5DEDA Combi-Blocks, Inc.

2-Benzofurancarboxaldehyde, 4,7-dimethyl-

  • MDL number MFCD13192460
  • Cas Number : 196519-54-7
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4446

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4446
Availability details for
Quantity
  • Availability
  • Add to Cart