COMH93D5DEE6 Combi-Blocks, Inc.

4-(4-Oxopiperidin-1-yl)benzamide

  • MDL number MFCD13192606
  • Cas Number : 340756-87-8
  • Molecular Weight : 218.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4459

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4459
Availability details for
Quantity
  • Availability
  • Add to Cart