COMH93D5DEEC Combi-Blocks, Inc.

tert-Butyl 4-(4-bromobenzoyl)-4'-methyl-[1,4'-bipiperidine]-1'-carboxylate

  • MDL number MFCD13192704
  • Cas Number : 305794-45-0
  • Molecular Weight : 465.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4465

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4465
Availability details for
Quantity
  • Availability
  • Add to Cart