COMH93D5DEF1 Combi-Blocks, Inc.

Benzene, 4-ethynyl-1-methoxy-2-(1-methylethoxy)-

  • MDL number MFCD13192740
  • Cas Number : 439585-86-1
  • Molecular Weight : 190.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4472

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4472
Availability details for
Quantity
  • Availability
  • Add to Cart