COMH93D5DF14 Combi-Blocks, Inc.

N1-(2,2-Dimethoxyethyl)ethane-1,2-diamine

  • MDL number MFCD13194098
  • Cas Number : 1343699-21-7
  • Molecular Weight : 148.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4510

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4510
Availability details for
Quantity
  • Availability
  • Add to Cart