COMH93D5DF6A Combi-Blocks, Inc.

4-Cyclohexylmethanesulfonylaniline

  • MDL number MFCD13471705
  • Cas Number : 1304343-74-5
  • Molecular Weight : 253.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4609

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4609
Availability details for
Quantity
  • Availability
  • Add to Cart