COMH93D5DF7B Combi-Blocks, Inc.

3-(4-Bromo-2-nitrophenoxy)azetidine

  • MDL number MFCD13559907
  • Cas Number : 1220027-84-8
  • Molecular Weight : 273.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4626

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4626
Availability details for
Quantity
  • Availability
  • Add to Cart