COMH93D5DFA9 Combi-Blocks, Inc.

N-Isopropylpiperidine-4-amine dihydrochloride

  • MDL number MFCD13689306
  • Cas Number : 870301-32-9
  • Molecular Weight : 215.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4682

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4682
Availability details for
Quantity
  • Availability
  • Add to Cart