COMH93D5DFC9 Combi-Blocks, Inc.

Methyl 2-cyano-3-oxo-3-phenylpropanoate

  • MDL number MFCD14281695
  • Cas Number : 3239-71-2
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4721

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4721
Availability details for
Quantity
  • Availability
  • Add to Cart