COMH93D5DFDD Combi-Blocks, Inc.

N-(2-Chloroacetyl)-4-fluorobenzamide

  • MDL number MFCD14551320
  • Cas Number : 897027-67-7
  • Molecular Weight : 215.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4746

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4746
Availability details for
Quantity
  • Availability
  • Add to Cart