COMH93D5E012 Combi-Blocks, Inc.

3-(4-Isoquinolinyl)benzaldehyde

  • MDL number MFCD14584528
  • Cas Number : 1023554-10-0
  • Molecular Weight : 233.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4811

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4811
Availability details for
Quantity
  • Availability
  • Add to Cart