COMH93D5E035 Combi-Blocks, Inc.

4-Chloro-6-(1h-1,2,4-triazol-1-yl)pyrimidine

  • MDL number MFCD14611280
  • Cas Number : 135948-75-3
  • Molecular Weight : 181.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4850

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4850
Availability details for
Quantity
  • Availability
  • Add to Cart