COMH93D5E03C Combi-Blocks, Inc.

Methyl 2-amino-3-(4-methoxyphenyl)propanoate

  • MDL number MFCD14618795
  • Cas Number : 61780-18-5
  • Molecular Weight : 209.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4858

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4858
Availability details for
Quantity
  • Availability
  • Add to Cart