COMH93D5E058 Combi-Blocks, Inc.

tert-Butyl 2-(6-bromo-1h-indol-3-yl)ethylcarbamate

  • MDL number MFCD14635678
  • Cas Number : 209168-88-7
  • Molecular Weight : 339.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4894

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4894
Availability details for
Quantity
  • Availability
  • Add to Cart