COMH93D5E061 Combi-Blocks, Inc.

tert-Butyl (1s,4s,5r)-rel-5-amino-2-azabicyclo[2.2.1]heptane-2-carboxylate

  • MDL number MFCD14636634
  • Cas Number : 1290539-90-0
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4903

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-4903
Availability details for
Quantity
  • Availability
  • Add to Cart