COMH93D5E0DE Combi-Blocks, Inc.

4-Iodo-6-methoxypyridin-3-ol

  • MDL number MFCD14705086
  • Cas Number : 431942-27-7
  • Molecular Weight : 251.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5039

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5039
Availability details for
Quantity
  • Availability
  • Add to Cart