COMH93D5E196 Combi-Blocks, Inc.

4-Methyl-6-(pyridin-3-yl)pyridin-3-amine

  • MDL number MFCD15475147
  • Cas Number : 1214379-41-5
  • Molecular Weight : 185.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5243

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5243
Availability details for
Quantity
  • Availability
  • Add to Cart