COMH93D5E21C Combi-Blocks, Inc.

tert-Butyl 2-oxo-decahydro-1,6-naphthyridine-6-carboxylate

  • MDL number MFCD16039664
  • Cas Number : 1221818-65-0
  • Molecular Weight : 254.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5404

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5404
Availability details for
Quantity
  • Availability
  • Add to Cart