COMH93D5E233 Combi-Blocks, Inc.

2-Amino-4-isopropylpyrimidine-5-carboxylic acid

  • MDL number MFCD16082809
  • Cas Number : 127958-03-6
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5429

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5429
Availability details for
Quantity
  • Availability
  • Add to Cart