COMH93D5E26A Combi-Blocks, Inc.

1-Pentylpiperidin-4-ol

  • MDL number MFCD16152965
  • Cas Number : 141430-49-1
  • Molecular Weight : 171.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5492

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5492
Availability details for
Quantity
  • Availability
  • Add to Cart