COMH93D5E272 Combi-Blocks, Inc.

(1-(Cyclopropylmethyl)piperidin-3-yl)methanol

  • MDL number MFCD16162801
  • Cas Number : 1247685-08-0
  • Molecular Weight : 169.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5502

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5502
Availability details for
Quantity
  • Availability
  • Add to Cart