COMH93D5E279 Combi-Blocks, Inc.

5-(4-(Trifluoromethyl)phenoxy)pentan-1-amine

  • MDL number MFCD16195852
  • Cas Number : 1274031-83-2
  • Molecular Weight : 247.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5511

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5511
Availability details for
Quantity
  • Availability
  • Add to Cart