COMH93D5E294 Combi-Blocks, Inc.

1,3-Oxazolidine

  • MDL number MFCD16251479
  • Cas Number : 504-76-7
  • Molecular Weight : 73.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5544

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5544
Availability details for
Quantity
  • Availability
  • Add to Cart