COMH93D5E296 Combi-Blocks, Inc.

4-Iodo-2-(trifluoromethyl)phenol

  • MDL number MFCD16251617
  • Cas Number : 1132942-88-1
  • Molecular Weight : 288.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5547

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5547
Availability details for
Quantity
  • Availability
  • Add to Cart