COMH93D5E2AD Combi-Blocks, Inc.

3-(Aminomethyl)-n-(propan-2-yl)pyridin-2-amine

  • MDL number MFCD16301188
  • Molecular Weight : 165.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5574

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5574
Availability details for
Quantity
  • Availability
  • Add to Cart