COMH93D5E2B0 Combi-Blocks, Inc.

3-(3,4-Difluorophenyl)prop-2-enal

  • MDL number MFCD16301257
  • Cas Number : 1334528-90-3
  • Molecular Weight : 168.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5577

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5577
Availability details for
Quantity
  • Availability
  • Add to Cart