COMH93D5E2B6 Combi-Blocks, Inc.

1-(2-Ethylphenyl)pyrrolidin-2-one

  • MDL number MFCD16302904
  • Cas Number : 1101046-65-4
  • Molecular Weight : 189.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5583

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5583
Availability details for
Quantity
  • Availability
  • Add to Cart