COMH93D5E2E3 Combi-Blocks, Inc.

Benzoxazole, 6-bromo-2-phenyl-

  • MDL number MFCD16468785
  • Cas Number : 537025-33-5
  • Molecular Weight : 274.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5633

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5633
Availability details for
Quantity
  • Availability
  • Add to Cart