COMH93D5E2FA Combi-Blocks, Inc.

1-(2-Phenoxyethyl)-1h-pyrazole-4-carboxylic acid

  • MDL number MFCD16497686
  • Cas Number : 1307601-91-7
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5661

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5661
Availability details for
Quantity
  • Availability
  • Add to Cart