COMH93D5E31C Combi-Blocks, Inc.

3-Bromo-5-fluoropyridine-2-carbaldehyde

  • MDL number MFCD16606511
  • Cas Number : 1227603-05-5
  • Molecular Weight : 204

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5703

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5703
Availability details for
Quantity
  • Availability
  • Add to Cart