COMH93D5E34D Combi-Blocks, Inc.

2-Fluoro-5-(trifluoromethyl)isonicotinic acid

  • MDL number MFCD16611302
  • Cas Number : 1227574-99-3
  • Molecular Weight : 209.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5768

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5768
Availability details for
Quantity
  • Availability
  • Add to Cart