COMH93D5E37C Combi-Blocks, Inc.

Benzyl 3-(tert-butoxycarbonyl)benzylcarbamate

  • MDL number MFCD16620767
  • Cas Number : 102638-44-8
  • Molecular Weight : 341.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5825

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5825
Availability details for
Quantity
  • Availability
  • Add to Cart