COMH93D5E380 Combi-Blocks, Inc.

tert-Butyl 4-(3-bromophenyl)-3-oxopiperazine-1-carboxylate

  • MDL number MFCD16620836
  • Cas Number : 873224-95-4
  • Molecular Weight : 355.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5829

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5829
Availability details for
Quantity
  • Availability
  • Add to Cart