COMH93D5E383 Combi-Blocks, Inc.

Nicotinamide riboside chloride

  • MDL number MFCD16621007
  • Cas Number : 1341-23-7
  • Molecular Weight : 255.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5832

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5832
Availability details for
Quantity
  • Availability
  • Add to Cart