COMH93D5E3E8 Combi-Blocks, Inc.

N-(Prop-2-en-1-yl)-2-(trifluoromethoxy)aniline

  • MDL number MFCD16684817
  • Molecular Weight : 217.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5952

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5952
Availability details for
Quantity
  • Availability
  • Add to Cart