COMH93D5E3E9 Combi-Blocks, Inc.

N-[(4-Fluoro-2-methylphenyl)methyl]cyclopropanamine

  • MDL number MFCD16686571
  • Cas Number : 1250981-88-4
  • Molecular Weight : 179.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5953

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5953
Availability details for
Quantity
  • Availability
  • Add to Cart