COMH93D5E3FC Combi-Blocks, Inc.

5-(Morpholin-4-yl)pentan-1-ol

  • MDL number MFCD16696205
  • Molecular Weight : 173.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5973

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5973
Availability details for
Quantity
  • Availability
  • Add to Cart