COMH93D5E400 Combi-Blocks, Inc.

1-(5-Chlorothiophene-2-carbonyl)piperazine

  • MDL number MFCD16699455
  • Cas Number : 864815-96-3
  • Molecular Weight : 230.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5977

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-5977
Availability details for
Quantity
  • Availability
  • Add to Cart